Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oil Red O Solution, 250 mL, G-Biosciences
SCI_ED

Catalog No. S001145
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001145 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001145 Supplier G-Biosciences Supplier No. 7862250
Add to Cart
Add to Cart

Oil Red O Solution is used for staining of lipid materials.

  • It imparts red-orange color to the lipids.

Specifications

CAS 1320-06-5
Molecular Weight (g/mol) 408.49
Grade Histological
Color Orange/Red
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.