Learn More
Description
Oil red O solution has been used to stain neutral lipids and adipocytes and to visualize fat droplets in fibroblasts.
Specifications
Specifications
| CAS | 1320-06-5 |
| Concentration | 0.5% in Isopropanol |
| Color | Dark Red |
| Quantity | 500 mL |
| Molecular Weight (g/mol) | 408.49 g/mol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.