Learn More
Scytek Laboratories OIL RED O STAIN KIT
SDP
Catalog No. NC0776970
Encompass_Preferred
Oil Red O Stain Kit (For Fat) is intended for use in the histological visualization of fat cells and neutral fat. This kit may be used ONLY on frozen tissue sections, fresh smears, or touch preps. Fat Cells: Red Neutral Fat: Red Nuclei: Blue
Catalog No. NC0776970 Supplier Scytek Laboratories Supplier No. ORK1
Oil Red O Stain Kit (For Fat) is intended for use in the histological visualization of fat cells and neutral fat. This kit may be used ONLY on frozen tissue sections, fresh smears, or touch preps. Fat Cells: Red Neutral Fat: Red Nuclei: Blue