Learn More
Newcomer Supply Oil Red O Stain, Propylene Glycol, 500 ml
SDP
Catalog No. NC2034720
Encompass
Newcomer Supply Oil Red O Stain, Propylene Glycol procedure is classified as a physical staining method and is used for identification of fat/lipid in frozen sections. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
This item is not returnable. View return policy
Catalog No. NC2034720 Supplier Newcomer Supply Supplier No. 12772B
Newcomer Supply Oil Red O Stain, Propylene Glycol procedure is classified as a physical staining method and is used for identification of fat/lipid in frozen sections. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.