Learn More
Newcomer Supply Oil Red O Stain, Isopropanol, 250 ml
SDP
Catalog No. NC1443713
Encompass
Newcomer Supply Oil Red O Stain, Isopropanol procedure, classified as a physical staining method, is used for identification of fat/lipid in frozen sections. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
Catalog No. NC1443713 Supplier Newcomer Supply Supplier No. 1277A
Newcomer Supply Oil Red O Stain, Isopropanol procedure, classified as a physical staining method, is used for identification of fat/lipid in frozen sections. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.