Learn More
MSC Clift Industries, Inc. Oil Stain Remover, Oil Vanish, 55 Gallon, Liquid
Catalog No. 502837254
Encompass
Oil Vanish Oil Stain Remover is a potent solution designed to effectively eliminate oil and grease stains from various surfaces. It is ideal for use on concrete, driveways, garages, and parking areas, offering a clear liquid formulation for robust cleaning.
  • Effectively removes oil and grease stains from concrete and masonry surfaces.
  • Suitable for driveways, garages, parking areas, fueling areas, and manufacturing facilities.
  • Tackles specific stains like motor oil, anti-freeze, brake fluid, diesel fuel, and tar.
  • Features a clear liquid, odorless, and non-flammable formula.
  • Neutral pH of 7.0.
  • Zero VOC content.
Catalog No. 50-283-7254 Supplier MSC Supplier No. 48380091
Oil Vanish Oil Stain Remover is a potent solution designed to effectively eliminate oil and grease stains from various surfaces. It is ideal for use on concrete, driveways, garages, and parking areas, offering a clear liquid formulation for robust cleaning.
  • Effectively removes oil and grease stains from concrete and masonry surfaces.
  • Suitable for driveways, garages, parking areas, fueling areas, and manufacturing facilities.
  • Tackles specific stains like motor oil, anti-freeze, brake fluid, diesel fuel, and tar.
  • Features a clear liquid, odorless, and non-flammable formula.
  • Neutral pH of 7.0.
  • Zero VOC content.