Learn More
CTL Scientific Supply Corp OIL TEST PAPER
SDP
Catalog No. 50300305 Shop All CTL Scientific Supply Products
Encompass_Preferred
OIL TEST PAPER 100 strips/pack. This light blue paper turns dark blue in the presence of hydrocarbons, particularly gasoline, fuel oil, lubricating oil, etc.
Catalog No. 50-300-305 Supplier CTL Scientific Supply Corp Supplier No. 90760
OIL TEST PAPER 100 strips/pack. This light blue paper turns dark blue in the presence of hydrocarbons, particularly gasoline, fuel oil, lubricating oil, etc.