Learn More
3B Scientific™ Oil Test Paper
SDPSCI_ED
Catalog No. S20741 Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
1
Catalog No. Quantity
S20741 1
1 options
Catalog No. S20741 Supplier 3B Scientific™ Supplier No. W11706
Add to Cart
Add to Cart
3B Scientific™ Oil-Test Paper allows users to test for oil in water or soil and to show hydrocarbons, especially in fuel (Diesel) and motor oil
  • Even if the water is self-coloured, the deep blue colouring of the test tape follows while having small oil contents
  • Package with 100 tapes 20 x 70mm
  • Description in English and German
Specifications
Product Type Biology Supplies
Includes 100 Tapes
Width (Metric) 5 cm
Height (Metric) 2.5 cm
Quantity 1
Material Paper, Plastic
Color White, Blue
Languages English