Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry Oil of turpentine | 2L | 8006-64-2 | MFCD00147790

Catalog No. 2424525L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred

CAS #: 8006-64-2
MDL #: MFCD00147790
UNSPSC Code: 12352103

Catalog No. 24-245-25L Supplier Sigma Organic Chemistry Supplier No. 242452.5L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS #: 8006-64-2
MDL #: MFCD00147790
UNSPSC Code: 12352103

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.