Learn More
Chemglass Life Sciences Oil, Vacuum Pump, Duo-Seal, 5 Gallon
SDP
Catalog No. 50872677
Encompass_Preferred
Oil, Vacuum Pump, Duo-Seal, 5 Gallon. Oil is specially formulated for use in either belt driven or direct drive vacuum pumps (AF-0349 or AF-0350). Oil is available in three types, Duo-Seal, Premium and Gold. The Duo-Seal oil is for use with belt driven (low RPM) AF-0350 pumps.
Catalog No. 50-872-677 Supplier Chemglass Life Sciences Supplier No. AF036012
Oil, Vacuum Pump, Duo-Seal, 5 Gallon. Oil is specially formulated for use in either belt driven or direct drive vacuum pumps (AF-0349 or AF-0350). Oil is available in three types, Duo-Seal, Premium and Gold. The Duo-Seal oil is for use with belt driven (low RPM) AF-0350 pumps.