Learn More
Chemglass Life Sciences Oil, Vacuum Pump, Gold Oil, Liter
SDP
Catalog No. 50872681
Encompass_Preferred
Oil, Vacuum Pump, Gold Oil, Liter. Oil is specially formulated for use in either belt driven or direct drive vacuum pumps (AF-0349 or AF-0350). Oil is available in three types, Duo-Seal, Premium and Gold. The Gold oil is a double distilled synthetic hydrocarbon oil that has no aromatic compounds or sulfur. Gold oil will offer longer life and superior protection for the internal components in corrosive pumping applications.
Catalog No. 50-872-681 Supplier Chemglass Life Sciences Supplier No. AF036016
Oil, Vacuum Pump, Gold Oil, Liter. Oil is specially formulated for use in either belt driven or direct drive vacuum pumps (AF-0349 or AF-0350). Oil is available in three types, Duo-Seal, Premium and Gold. The Gold oil is a double distilled synthetic hydrocarbon oil that has no aromatic compounds or sulfur. Gold oil will offer longer life and superior protection for the internal components in corrosive pumping applications.