Learn More
Chemglass Life Sciences Oil, Vacuum Pump, Premium Oil, Liter
SDP
Catalog No. 50872678
Encompass_Preferred
Oil, Vacuum Pump, Premium Oil, Liter. Oil is specially formulated for use in either belt driven or direct drive vacuum pumps (AF-0349 or AF-0350). Oil is available in three types, Duo-Seal, Premium and Gold. The Premium oil is a triple distilled hydrocarbon oil resistant to breakdown at the high RPM of direct drive pumps.
Catalog No. 50-872-678 Supplier Chemglass Life Sciences Supplier No. AF036013
Oil, Vacuum Pump, Premium Oil, Liter. Oil is specially formulated for use in either belt driven or direct drive vacuum pumps (AF-0349 or AF-0350). Oil is available in three types, Duo-Seal, Premium and Gold. The Premium oil is a triple distilled hydrocarbon oil resistant to breakdown at the high RPM of direct drive pumps.