Learn More
MSC Karcher Hose, PU, DN40, Oil-Resistant, Electroconductive, 1 ea
Catalog No. 502819161
Encompass
This hose is an oil-resistant, electroconductive PU hose with a DN 40 diameter, suitable for abrasive suction media and capable of withstanding temperatures up to 80 °C. It has a length of 5 meters.
  • Oil-resistant
  • Electroconductive
  • Suitable for abrasive suction media
  • Temperature-resistant up to 80 °C
Catalog No. 50-281-9161 Supplier MSC Supplier No. 16616211
This hose is an oil-resistant, electroconductive PU hose with a DN 40 diameter, suitable for abrasive suction media and capable of withstanding temperatures up to 80 °C. It has a length of 5 meters.
  • Oil-resistant
  • Electroconductive
  • Suitable for abrasive suction media
  • Temperature-resistant up to 80 °C