Learn More
MSC Nilfisk Sump Pump Vacuum, Electric, 19.8 gal, Stainless Steel
Catalog No. 502839097
Encompass
This industrial vacuum cleaner is engineered for the metalworking industry, efficiently collecting metal chips, oily cuttings, lubricants, and coolant spills. It separates liquids from solids and facilitates quick discharge, making it ideal for maintaining a clean and efficient workspace. Its robust construction ensures durability in demanding environments.
  • Collects and separates mixed liquids and solids for recycling
  • Diverting valve allows for quick tank emptying
  • Tilting motor head provides easy access to the internal chip basket
  • Constructed from 304 stainless steel for durability
  • Features a removable collection bin and automatic shut-off float system
  • Internal oil mist filter protects the motor from oil residue
  • Transparent liquid level sight tube for easy monitoring
  • Small and maneuverable design for use in tight spaces
Catalog No. 50-283-9097 Supplier MSC Supplier No. 35210368
Add to Cart
Add to Cart
Request a Quote
This industrial vacuum cleaner is engineered for the metalworking industry, efficiently collecting metal chips, oily cuttings, lubricants, and coolant spills. It separates liquids from solids and facilitates quick discharge, making it ideal for maintaining a clean and efficient workspace. Its robust construction ensures durability in demanding environments.
  • Collects and separates mixed liquids and solids for recycling
  • Diverting valve allows for quick tank emptying
  • Tilting motor head provides easy access to the internal chip basket
  • Constructed from 304 stainless steel for durability
  • Features a removable collection bin and automatic shut-off float system
  • Internal oil mist filter protects the motor from oil residue
  • Transparent liquid level sight tube for easy monitoring
  • Small and maneuverable design for use in tight spaces