Learn More
Perkin Elmer US LLC OilExpress 4 Installation Kit
Catalog No. 502104240
Encompass_Preferred
This kit includes Part Number Description Material Volume N9308350 OilExpress 4 FTIR Control Standard 15W40 Diesel Oil 100 mL N9308351 OilExpress 4 Test Oil 120cSt PAO Oils 500 mL N9308352 OilExpress 4 Test Oil 360cSt PAO Oils 100 mL N9308354 OilExpress 4 System Liquid Detergent Additive N/A 100 mL N9308377 OilExpress4 Calibration Fluid Squalane (99% pure) 100 mL
Catalog No. 50-210-4240 Supplier Perkin Elmer US LLC Supplier No. N9300850
This kit includes Part Number Description Material Volume N9308350 OilExpress 4 FTIR Control Standard 15W40 Diesel Oil 100 mL N9308351 OilExpress 4 Test Oil 120cSt PAO Oils 500 mL N9308352 OilExpress 4 Test Oil 360cSt PAO Oils 100 mL N9308354 OilExpress 4 System Liquid Detergent Additive N/A 100 mL N9308377 OilExpress4 Calibration Fluid Squalane (99% pure) 100 mL