Learn More
Cole-Parmer JUN-AIR Jun-Air Oilless Piston Air Compressor w/ Dryer & Sound Proof Cabinet, 4 L, 230V
Catalog No. NC1400046 Shop All Cole-Parmer Products
Encompass
These oil-free air compressors provide air power suitable for laboratories and other environments with delicate machinery and critical instrumentation. Choose a model with a protective metal cabinet housing to reduce the sound level even further than the base model's already quiet level. For applications requiring dry air, such as those sensitive to microbial growth, choose from the models with the integrated adsorbent dryer.
Catalog No. NC1400046 Supplier Cole-Parmer Supplier No. 1620225
Add to Cart
Add to Cart
Request a Quote
These oil-free air compressors provide air power suitable for laboratories and other environments with delicate machinery and critical instrumentation. Choose a model with a protective metal cabinet housing to reduce the sound level even further than the base model's already quiet level. For applications requiring dry air, such as those sensitive to microbial growth, choose from the models with the integrated adsorbent dryer.