Learn More
Uline Oily Waste Can - Red, 6 Gallon
Catalog No. NC2865749
Encompass
Safe, secure disposal for oily and solvent-soaked rags.Prevents fires due to spontaneous combustion.Hands-free; foot pedal opens lid.Lead-free galvanized steel with powder-coat finish
Catalog No. NC2865749 Supplier Uline Supplier No. H1846R
Safe, secure disposal for oily and solvent-soaked rags.Prevents fires due to spontaneous combustion.Hands-free; foot pedal opens lid.Lead-free galvanized steel with powder-coat finish