Learn More
Justrite Oily Waste Cans
Catalog No. 19085900 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (English):
21 gal.
Catalog No. Capacity (English)
19-085-900 21 gal.
1 options
This item is not returnable. View return policy
Catalog No. 19-085-900 Supplier Justrite Supplier No. 09701
Add to Cart
Add to Cart
This item is not returnable. View return policy
Dispose of oil-soaked rags and prevent fires caused by stray sparks or spontaneous combustion.
  • Lid stays closed when not in use to isolate contents from fire sources and limit available oxygen
  • Round construction and elevated base allow circulation of air around can to disperse heat and reduce moisture build-up and rusting
  • Durable steel construction with a powder-coat finish provides chemical resistance
  • Swinging carry handle for portability
  • Warning label with pictographs reduces miscommunication
  • FM-approved
  • Meets OSHA requirements
  • Foot-operated cans meet TÜV certification requirements
Specifications
Capacity (English) 21 gal.
Color Yellow
Material Galvanized Steel
Product Type Oily Waste Safety Can (Foot Lever)