Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Ointment Tube Pliers

Catalog No. 21124922 Shop All Surgical Design Products
Change view
Click to view available options
Handle Material:
PVC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Handle Material
21-124-922 PVC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-124-922 Supplier Surgical Design™ Supplier No. OP07
Add to Cart
Add to Cart

Ointment tube pliers help squeeze, flatten, and empty ointment, cream, or gel tubes efficiently with minimal waste using a durable, easy-grip design.

  • Optimized for metal and plastic tubes—ideal for ointments, gels, creams, and pastes
  • Provides strong, even pressure to extract contents fully and reduce product waste
  • Ergonomic handle design ensures comfortable, non-slip grip during use
  • Smooth compression action prevents tube tearing or puncturing
  • Durable stainless-steel construction suitable for medical, lab, and general-purpose use
  • Compact and lightweight for easy storage in kits, clinics, or workstations

Specifications

Product Type Crimping Pliers
For Use With (Equipment) Ointment Tube
Grade Standard
Handle Material PVC
Material Stainless Steel
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.