Learn More
Chiral Technologies, Inc. OJ-H 5UM 250X4.6
Catalog No. NC9819616
Encompass
Daicel's High Performance 'H' series Chiral Columns, 5micrometer Particle Size, 4.6mm Internal Diameter, 250mm Column Length, Cellulose tris (4-methylbenzoate) Adsorbent
Catalog No. NC9819616 Supplier Chiral Technologies, Inc. Supplier No. 17325
Daicel's High Performance 'H' series Chiral Columns, 5micrometer Particle Size, 4.6mm Internal Diameter, 250mm Column Length, Cellulose tris (4-methylbenzoate) Adsorbent