Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OK-1™ Full Finger Anti-Vibration Pigskin Leather Gloves

Catalog No. 19816342
Change view
Click to view available options
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-816-342 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-816-342 Supplier OK-1™ Supplier No. OK995EM

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are constructed from premium grain pigskin leather with built-in elastic wrist support.

  • Premium grain pigskin leather
  • Strech nylon back for comfort
  • Built-in elastic wrist support
  • Hook and loop closure
  • Machine washable

Specifications

Cuff Material Elastic
Color Black
Hand Specific
Material Grain Pigskin Leather
Product Type Anti-vibration Gloves
Size Medium
For Use With (Application) Construction, Machine Operation, and Manufacturing
Glove Cuff Style Open
Series OK995E
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.