Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OK-1™ Half-Finger Anti-Vibration Gloves

Catalog No. 19821753
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-821-753 Medium
19-821-752 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-821-753 Supplier OK-1™ Supplier No. OK980M

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves feature a pre-curved design for worn-in comfort from the first use. Anti-vibration technology protects hands by absorbing and dissipating vibrations caused by the constant use of power tools.

  • A precurved shape and half finger design provide dexterity
  • Gloves offer superior protection from sources of vibration and impact
  • Maintain a consistent grip due to the curved design that forms to the natural shape of the hand
  • Machine washable

Specifications

Color Brown
Hand Specific
Material Grain Cowhide Leather
Product Type Work Gloves
Size Medium
For Use With (Application) Construction, Machine Operation, and Manufacturing
Series OK980
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.