Learn More
ATCC OK; Kidney Proximal Tubule; Virginia opossum (Didelphis virginiana)
Catalog No. 502382700
Encompass
OK is an epithelial cell line that was isolated from the proximal tubule of an adult female opossum. This cell line was deposited by MM Miller.
Catalog No. 50-238-2700 Supplier ATCC Supplier No. CRL1840
Add to Cart
Add to Cart
OK is an epithelial cell line that was isolated from the proximal tubule of an adult female opossum. This cell line was deposited by MM Miller.