Learn More
Cayman Chemical Okadaic Acid, 78111-17-8, 100 µg
SDP
Supplier:  Cayman Chemical 10011490100
Encompass_Preferred
A potent inhibitor of certain serine/threonine protein phosphatases, targeting the multiple isoforms of PP1 (IC50 = 10-50 nM), both isoforms of PP2A (IC50 = 0.5 nM) and PP3 (IC50 = 4 nM); a very weak inhibitor of PP2B (IC50 > 2 μM); does not inhibit PP2C or other phosphatases
Catalog No. NC1405924