Learn More
Enzo Life Sciences Okadaic acid . ammonium salt (high purity), (1mg), CAS Number: 155716-06-6
SDP
Supplier:  Enzo Life Sciences ALX350010M001
Encompass_Preferred
Inhibitor of PP1 and PP2A. Salt form of okadaic acid (Prod. No. ALX-350-003), with slightly greater stability than the free acid after it is put into stock solution (in organic solvents). Potent inhibitor of protein phosphatase 1 (PP1) and 2A (PP2A). Potent tumor promotor and neurotoxin. Alternative Name: Halochondrine A . NH4, 9,10-Deepithio-9,10-didehydroacanthifolicin. NH4. Formula: C44H67O13 . NH4. MW: 804.0 . 18.0. CAS Number: 155716-06-6. Purity: ≥98% (TLC). Appearance: White to off-white solid. Solubility: Soluble in ethanol or methanol. Long Term Storage: -20°C
Catalog No. NC0953616