Learn More
Enzo Life Sciences Okadaic acid (high purity), (100µg), CAS Number: 78111-17-8
SDP
Catalog No. NC1223108
Encompass_Preferred
Inhibitor of PP1 and PP2A. Alternative Name: Halochondrine A, 9,10-Deepithio-9,10-didehydroacanthifolicin. Formula: C44H68O13. MW: 805.0. CAS Number: 78111-17-8. MI: 14: 6819. Purity: ≥98% (TLC). Appearance: White crystalline solid. Solubility: Soluble in chloroform, ethanol, methanol, acetone or ethyl acetate. Use/Stability: Use only fresh solutions. Long Term Storage: -20°C
Catalog No. NC1223108 Supplier Enzo Life Sciences Supplier No. ALX350003C100
Inhibitor of PP1 and PP2A. Alternative Name: Halochondrine A, 9,10-Deepithio-9,10-didehydroacanthifolicin. Formula: C44H68O13. MW: 805.0. CAS Number: 78111-17-8. MI: 14: 6819. Purity: ≥98% (TLC). Appearance: White crystalline solid. Solubility: Soluble in chloroform, ethanol, methanol, acetone or ethyl acetate. Use/Stability: Use only fresh solutions. Long Term Storage: -20°C