Learn More
Medchemexpress LLC HY-N6673 5mg Medchemexpress, Okanin CAS:484-76-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN66735MG
Encompass_Preferred
Medchemexpress, HY-N6673 5mg Okanin CAS:484-76-4 Okanin, effective constituent of the flower tea Coreopsis tinctoria, attenuates LPS-induced microglial activation through inhibition of the TLR4/NF-κB signaling pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2955