Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Public Seating™ Oklahoma Sound™ Graystone Lectern
SCI_ED

Catalog No. S83744 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Material:
Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S83744 Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S83744 Supplier National Public Seating™ Supplier No. GSL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lectern for teachers with storage space

  • Designer curved gray plywood
  • Includes:
    • Lectern
    • Large reading surface
    • Two side pull-out shelves
    • Sliding keyboard tray
    • Cabinet with three large shelves
    • Oversized front drawer for storage
    • 2.5 in. twin wheel matching casters

Specifications

Product Type Lectern
Color Charcoal
Dimensions (L x W) Reading surface: 22.75 x 18.25 in., Side pull-out shelves: 14.25 x 12 in., Keyboard tray: 17.5 x 9.25 in.
For Use With (Application) For Lectures
Warranty 10 years
Material Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.