Learn More
Sciencell Research Laboratories Oral Keratinocyte Medium, 500 ml, Sterile, Liquid, pH 7.4, HEPES and bicarbonate buffered, Selective support for oral keratinocytes, Serum-free, Research Use Only
Catalog No. NC9714660
Encompass
Oral Keratinocyte Medium, 500 ml, Sterile, Liquid, pH 7.4, HEPES and bicarbonate buffered, Selective support for oral keratinocytes, Serum-free, Research Use Only
Catalog No. NC9714660 Supplier Sciencell Research Laboratories Supplier No. 2611
Oral Keratinocyte Medium, 500 ml, Sterile, Liquid, pH 7.4, HEPES and bicarbonate buffered, Selective support for oral keratinocytes, Serum-free, Research Use Only