Learn More
Aviva Systems Biology Corp OLAH Antibody - N-terminal region (ARP56264_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56264_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OLAH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human OLAH. Alternate names: AURA1, FLJ11106, MGC51852, SAST, THEDC1ApplicationWB. Species reactivity: Guinea Pig, Human, Rat
Catalog No. 50-014-61787