Learn More
Medchemexpress LLC Olanexidine (hydrochloride) | 146509-94-6 | MFCD32201949 | 96.3% | 408.80 | C17H28Cl3N5 | 25MG
SDP
Catalog No. 5000237283
Encompass_Preferred
Olanexidine (hydrochloride) is a monobiguanide antibacterial research chemical with bactericidal activity that disrupts bacterial cell membranes. It is supplied as a hydrochloride salt for laboratory research and is provided with batch COA and SDS documentation confirming identity, formula, molecular weight, and purity.
  • Bactericidal activity against gram-positive and gram-negative bacteria.
  • Available in small research pack sizes, such as 25 mg, for laboratory studies.
  • Includes certificate of analysis and safety data sheet for verification.
  • Purity reported at approximately 96.3%.
  • Supplied as a hydrochloride salt for improved stability and handling.
Catalog No. 50-002-37283 Supplier Medchemexpress LLC Supplier No. HY125654A25MG
Olanexidine (hydrochloride) is a monobiguanide antibacterial research chemical with bactericidal activity that disrupts bacterial cell membranes. It is supplied as a hydrochloride salt for laboratory research and is provided with batch COA and SDS documentation confirming identity, formula, molecular weight, and purity.
  • Bactericidal activity against gram-positive and gram-negative bacteria.
  • Available in small research pack sizes, such as 25 mg, for laboratory studies.
  • Includes certificate of analysis and safety data sheet for verification.
  • Purity reported at approximately 96.3%.
  • Supplied as a hydrochloride salt for improved stability and handling.