Learn More
Medchemexpress LLC Olanexidine (hydrochloride) | 146509-94-6 | MFCD32201949 | 96.3% | 408.80 g/mol | C17H28Cl3N5 | 100MG
SDP
Catalog No. 5000236067
Encompass_Preferred
Olanexidine (hydrochloride) is a monobiguanide antibacterial small molecule for research use. It exhibits broad-spectrum bactericidal activity against both Gram-positive and Gram-negative bacteria and is supplied as the hydrochloride salt with characterized purity and molecular properties.
Catalog No. 50-002-36067 Supplier Medchemexpress LLC Supplier No. HY125654A100MG
Olanexidine (hydrochloride) is a monobiguanide antibacterial small molecule for research use. It exhibits broad-spectrum bactericidal activity against both Gram-positive and Gram-negative bacteria and is supplied as the hydrochloride salt with characterized purity and molecular properties.