Learn More
Medchemexpress LLC Olanexidine (hydrochloride) | 146509-94-6 | MFCD32201949 | 96.3% | 408.80 g/mol | C17H28Cl3N5 | 10MM 1ML
SDP
Catalog No. 5000236069
Encompass_Preferred
Olanexidine hydrochloride is a monobiguanide antibacterial agent active against a broad range of Gram-positive and Gram-negative bacteria. It is supplied for research use and offered as both a solid and a ready-to-use 10 mM solution in DMSO (1 mL). Typical applications include in vitro bactericidal testing and studies of antimicrobial mechanism; identity and purity are documented by SDS and batch COA.
Catalog No. 50-002-36069 Supplier Medchemexpress LLC Supplier No. HY125654A10MM 1ML
Olanexidine hydrochloride is a monobiguanide antibacterial agent active against a broad range of Gram-positive and Gram-negative bacteria. It is supplied for research use and offered as both a solid and a ready-to-use 10 mM solution in DMSO (1 mL). Typical applications include in vitro bactericidal testing and studies of antimicrobial mechanism; identity and purity are documented by SDS and batch COA.