Learn More
Medchemexpress LLC Olanzapine | 132539-06-1 | 99.9% | C17H20N4S | 50 MG
SDP
Catalog No. 5000353953
Encompass_Preferred
Olanzapine (Standard) is an analytical standard intended for research and analytical applications. It is an orally active, selective monoaminergic antagonist with high affinity for serotonin H1, 5HT2A/2C, 5HT3, 5HT6, dopamine D1-4, muscarinic M1-5, and adrenergic α1 receptors. Classified as an atypical antipsychotic, this compound serves as a reference standard for various assay methods.
  • Reference standard supplied assay
  • Used in qualitative, quantitative, and methodological research
  • Applications in HPLC, GC, and MS
  • Selective monoaminergic antagonist
  • High affinity for various serotonin, dopamine, muscarinic, and adrenergic receptors
Catalog No. 50-003-53953 Supplier Medchemexpress LLC Supplier No. HY14541R50MG
Olanzapine (Standard) is an analytical standard intended for research and analytical applications. It is an orally active, selective monoaminergic antagonist with high affinity for serotonin H1, 5HT2A/2C, 5HT3, 5HT6, dopamine D1-4, muscarinic M1-5, and adrenergic α1 receptors. Classified as an atypical antipsychotic, this compound serves as a reference standard for various assay methods.
  • Reference standard supplied assay
  • Used in qualitative, quantitative, and methodological research
  • Applications in HPLC, GC, and MS
  • Selective monoaminergic antagonist
  • High affinity for various serotonin, dopamine, muscarinic, and adrenergic receptors