Learn More
Medchemexpress LLC Olanzapine | 132539-06-1 | MFCD00078044 | 99.96% | C17H20N4S | 250 MG
SDP
Catalog No. 5000352823
Encompass_Preferred
Olanzapine is a selective, orally active monoaminergic antagonist with high affinity binding to serotonin H1, 5HT2A/2C, 5HT3, 5HT6, dopamine D1-4, muscarinic M1-5, and adrenergic α1 receptors. It is classified as an atypical antipsychotic.
  • Selective monoaminergic antagonist
  • High affinity binding to multiple serotonin, dopamine, muscarinic, and adrenergic receptors
  • Atypical antipsychotic
Catalog No. 50-003-52823 Supplier Medchemexpress LLC Supplier No. 250MGHY14541
Olanzapine is a selective, orally active monoaminergic antagonist with high affinity binding to serotonin H1, 5HT2A/2C, 5HT3, 5HT6, dopamine D1-4, muscarinic M1-5, and adrenergic α1 receptors. It is classified as an atypical antipsychotic.
  • Selective monoaminergic antagonist
  • High affinity binding to multiple serotonin, dopamine, muscarinic, and adrenergic receptors
  • Atypical antipsychotic