Learn More
Medchemexpress LLC Olanzapine-d3 | 786686-79-1 | 98.6% | C17H17D3N4S | 1 MG
SDP
Catalog No. 5000356255
Encompass_Preferred
Olanzapine-d3 is the deuterium-labeled version of Olanzapine, a selective, orally active monoaminergic antagonist. It binds with high affinity to serotonin H1, 5HT2A/2C, 5HT3, 5HT6, dopamine D1-4, muscarinic M1-5, and adrenergic α1 receptors. Olanzapine is classified as an atypical antipsychotic, and its deuterated form can serve as a tracer.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-56255 Supplier Medchemexpress LLC Supplier No. HY14541S1MG
Add to Cart
Add to Cart
Olanzapine-d3 is the deuterium-labeled version of Olanzapine, a selective, orally active monoaminergic antagonist. It binds with high affinity to serotonin H1, 5HT2A/2C, 5HT3, 5HT6, dopamine D1-4, muscarinic M1-5, and adrenergic α1 receptors. Olanzapine is classified as an atypical antipsychotic, and its deuterated form can serve as a tracer.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated into drug molecules as tracers for quantitation.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.