Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Olanzapine-lactam | 221176-49-4 | 99.1% | 230.29 | 100 MG
SDP

Catalog No. 5000364955
Encompass_Preferred

Olanzapine-lactam (LY301664) is a metabolite of Olanzapine. It is a solid, light yellow to yellow in appearance.

  • Purity: 99.12%
  • Molecular weight: 230.29
  • Formula: C12H10N2OS
  • Solubility in DMSO: 100 mg/mL
  • Available in sizes from 5 mg to 10 g

Catalog No. 50-003-64955 Supplier Medchemexpress LLC Supplier No. 100MGHYW004412
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Olanzapine-lactam (LY301664) is a metabolite of Olanzapine. It is a solid, light yellow to yellow in appearance.

  • Purity: 99.12%
  • Molecular weight: 230.29
  • Formula: C12H10N2OS
  • Solubility in DMSO: 100 mg/mL
  • Available in sizes from 5 mg to 10 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.