Learn More
Medchemexpress LLC Olanzapine-lactam | 221176-49-4 | MFCD18252448 | 99.1% | 230.29 | 250 MG
SDP
Catalog No. 5000366222
Encompass_Preferred
Olanzapine-lactam (LY301664) is a metabolite of Olanzapine. It is intended for research use only.
  • Powder stable for up to 3 years at -20°C or 2 years at 4°C.
  • Solution stable for up to 6 months at -80°C or 1 month at -20°C.
  • Soluble in DMSO at 100 mg/mL.
  • Soluble in a 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline solution at ≥ 2.5 mg/mL.
Catalog No. 50-003-66222 Supplier Medchemexpress LLC Supplier No. 250MGHYW004412
Olanzapine-lactam (LY301664) is a metabolite of Olanzapine. It is intended for research use only.
  • Powder stable for up to 3 years at -20°C or 2 years at 4°C.
  • Solution stable for up to 6 months at -80°C or 1 month at -20°C.
  • Soluble in DMSO at 100 mg/mL.
  • Soluble in a 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline solution at ≥ 2.5 mg/mL.