Learn More
American Radiolabeled Chemicals Inc OLANZAPINE ME 14C 50UC
SDP
Catalog No. 50901272
Encompass_Preferred
Olanzapine-methyl-14C 50uCi in ethanol, 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 312.45, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-272 Supplier American Radiolabeled Chemicals Inc Supplier No. ARC 346350UC
Olanzapine-methyl-14C 50uCi in ethanol, 0.1 mCi/ml, S.A. 50-60 mCi/mmol, M.W. 312.45, Shipped in Dry Ice, Exclusive