Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Olanzapine N-oxide | 174794-02-6 | 98.0% | 100 MG
SDP

Catalog No. 5000233152
Encompass_Preferred

Olanzapine N-oxide is a research-grade metabolite of olanzapine supplied as a purified solid for laboratory research and analytical applications. It is intended for in vitro and in vivo metabolism studies and as an analytical reference; not for human use.

  • Purity 98.0%.
  • Molecular weight 328.43 g/mol.
  • Chemical formula C17H20N4OS.
  • Available in multiple package sizes, including 100 mg.
  • Used as an analytical reference standard for LC-MS and method development.
  • Intended for research use only; not for clinical or therapeutic use.

Catalog No. 50-002-33152 Supplier Medchemexpress LLC Supplier No. HY119393100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Olanzapine N-oxide is a research-grade metabolite of olanzapine supplied as a purified solid for laboratory research and analytical applications. It is intended for in vitro and in vivo metabolism studies and as an analytical reference; not for human use.

  • Purity 98.0%.
  • Molecular weight 328.43 g/mol.
  • Chemical formula C17H20N4OS.
  • Available in multiple package sizes, including 100 mg.
  • Used as an analytical reference standard for LC-MS and method development.
  • Intended for research use only; not for clinical or therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.