Learn More
Medchemexpress LLC Olanzapine N-oxide | 174794-02-6 | 98.0% | 50 MG
SDP
Catalog No. 5000233519
Encompass_Preferred
Olanzapine N-oxide is the N-oxide metabolite of olanzapine, provided as a research reference standard for metabolic and analytical studies.
  • High purity suitable for analytical standards.
  • Molecular formula C17H20N4OS, molecular weight 328.43 g·mol⁻¹.
  • Available in small benchtop quantities for research, such as 50 MG.
  • Intended for in vitro and in vivo metabolism research and analytical method development.
  • Packaged with supplier-reported purity and lot-specific documentation.
Catalog No. 50-002-33519 Supplier Medchemexpress LLC Supplier No. HY11939350MG
Olanzapine N-oxide is the N-oxide metabolite of olanzapine, provided as a research reference standard for metabolic and analytical studies.
  • High purity suitable for analytical standards.
  • Molecular formula C17H20N4OS, molecular weight 328.43 g·mol⁻¹.
  • Available in small benchtop quantities for research, such as 50 MG.
  • Intended for in vitro and in vivo metabolism research and analytical method development.
  • Packaged with supplier-reported purity and lot-specific documentation.