Learn More
Medchemexpress LLC Olanzapine N-oxide-d3 | 1189732-75-9 | 1 MG
SDP
Catalog No. 5000234160
Encompass_Preferred
Olanzapine N-Oxide-d3 is a deuterium-labeled reference standard of olanzapine N-oxide used as an internal standard or tracer in analytical, pharmacokinetic, and metabolite profiling studies.
  • Isotopically labeled (d3) internal standard for mass spectrometry
  • Supplied as a 1 mg vial for analytical use
  • Enables accurate quantification of olanzapine N-oxide and related metabolites
  • For research use only; not for human or clinical use
  • Deuterium labeling preserves chemical behavior while allowing mass distinction
Catalog No. 50-002-34160 Supplier Medchemexpress LLC Supplier No. HY119393S1MG
Olanzapine N-Oxide-d3 is a deuterium-labeled reference standard of olanzapine N-oxide used as an internal standard or tracer in analytical, pharmacokinetic, and metabolite profiling studies.
  • Isotopically labeled (d3) internal standard for mass spectrometry
  • Supplied as a 1 mg vial for analytical use
  • Enables accurate quantification of olanzapine N-oxide and related metabolites
  • For research use only; not for human or clinical use
  • Deuterium labeling preserves chemical behavior while allowing mass distinction