Learn More
Cayman Chemical Olanzapine-related Compound B
Supplier: Cayman Chemical 3378625 MG
A potential impurity found in commercial preparations of olanzapine; a degradation product formed by exposure to oxidative stress