Learn More
Cayman Chemical Olanzapine-related Compound B
SDP
Catalog No. 502948356
Encompass_Preferred
A potential impurity found in commercial preparations of olanzapine; a degradation product formed by exposure to oxidative stress
Catalog No. 50-294-8356 Supplier Cayman Chemical Supplier No. 3378610 MG
A potential impurity found in commercial preparations of olanzapine; a degradation product formed by exposure to oxidative stress