Learn More
Medchemexpress LLC HY-10162 100mg , Olaparib CAS:763113-22-0 Purity:>98%
SDP
Catalog No. 502258977
Encompass_Preferred
HY-10162 100mg Olaparib CAS: 763113-22-0 Olaparib (AZD2281; KU0059436) is a potent and orally active PARP inhibitor with IC50s of 5 and 1 nM for PARP1 and PARP2, respectively. Olaparib is an autophagy and mitophagy activator[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-8977 Supplier Medchemexpress LLC Supplier No. HY10162100MG
HY-10162 100mg Olaparib CAS: 763113-22-0 Olaparib (AZD2281; KU0059436) is a potent and orally active PARP inhibitor with IC50s of 5 and 1 nM for PARP1 and PARP2, respectively. Olaparib is an autophagy and mitophagy activator[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.