Learn More
Medchemexpress LLC HY-10162 50mg Medchemexpress, Olaparib CAS:763113-22-0 Purity:>98%
SDP
Catalog No. 502028673
Encompass_Preferred
Medchemexpress, HY-10162 50mg Olaparib CAS:763113-22-0 Olaparib (AZD2281; KU0059436) is a potent and orally active PARP inhibitor with IC50s of 5 and 1 nM for PARP1 and PARP2, respectively. Olaparib is an autophagy and mitophagy activator[2][3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8673 Supplier Medchemexpress LLC Supplier No. HY1016250MG
Medchemexpress, HY-10162 50mg Olaparib CAS:763113-22-0 Olaparib (AZD2281; KU0059436) is a potent and orally active PARP inhibitor with IC50s of 5 and 1 nM for PARP1 and PARP2, respectively. Olaparib is an autophagy and mitophagy activator[2][3][4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.