Learn More
Selleck Chemical LLC Olaparib (AZD2281) 100mg 763113-22-0 Ku-0059436
SDP
Catalog No. 508641
Encompass_Preferred
Olaparib (AZD2281, KU0059436) is a selective inhibitor of PARP1/2 with IC50 of 5 nM/1 nM in cell-free assays, 300-times less effective against tankyrase-1. Olaparib induces significant autophagy that is associated with mitophagy in cells with BRCA mutations. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-864-1 Supplier Selleck Chemical LLC Supplier No. S1060100MG
Olaparib (AZD2281, KU0059436) is a selective inhibitor of PARP1/2 with IC50 of 5 nM/1 nM in cell-free assays, 300-times less effective against tankyrase-1. Olaparib induces significant autophagy that is associated with mitophagy in cells with BRCA mutations. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.