Learn More
Medchemexpress LLC Olaparib (cyclopropylcarbonyl-d4) | 2143107-55-3 | 98.6% | 438.49 g·mol⁻¹ | C24H19D4FN4O3 | 10 MG
SDP
Catalog No. 5000249073
Encompass_Preferred
Olaparib-d4-1 is a deuterium-labeled isotopologue of Olaparib provided as a stable isotope reference standard for analytical and research use. It is designed for use as an internal standard in LC-MS/MS and quantitative assays, offering consistent labeling and documented characterization to support method development and validation.
  • Deuterium-labeled internal standard for mass spectrometry and quantitative assays.
  • High purity supports accurate quantitation (98.6%).
  • Documented molecular formula and molecular weight for precise calculations.
  • Supplied in a 10 mg vial suitable for trace-level analyses.
  • Applicable to pharmacokinetic, metabolite, and method validation studies.
Catalog No. 50-002-49073 Supplier Medchemexpress LLC Supplier No. HY10162S310MG
Olaparib-d4-1 is a deuterium-labeled isotopologue of Olaparib provided as a stable isotope reference standard for analytical and research use. It is designed for use as an internal standard in LC-MS/MS and quantitative assays, offering consistent labeling and documented characterization to support method development and validation.
  • Deuterium-labeled internal standard for mass spectrometry and quantitative assays.
  • High purity supports accurate quantitation (98.6%).
  • Documented molecular formula and molecular weight for precise calculations.
  • Supplied in a 10 mg vial suitable for trace-level analyses.
  • Applicable to pharmacokinetic, metabolite, and method validation studies.