Learn More
Medchemexpress LLC Olaparib-d4-1 | 2143107-55-3 | 98.6% | 438.49 g/mol | C24H19D4FN4O3 | 5 MG
SDP
Catalog No. 5000248688
Encompass_Preferred
Olaparib-d4-1 is a deuterium-labeled analogue of olaparib used as a stable-isotope reference standard for analytical and research applications. It provides a defined mass shift while preserving chemical behavior of the parent compound, enabling accurate quantification and tracer studies in mass spectrometry and related assays.
  • Deuterium-labeled reference standard for LC-MS analyses.
  • Molecular weight 438.49 g/mol with defined chemical formula.
  • High purity suitable for quantitative analytical workflows.
  • Offered in small measured packages for easy sample preparation.
  • Datasheet and safety data sheet available for handling and characterization.
Catalog No. 50-002-48688 Supplier Medchemexpress LLC Supplier No. HY10162S35MG
Olaparib-d4-1 is a deuterium-labeled analogue of olaparib used as a stable-isotope reference standard for analytical and research applications. It provides a defined mass shift while preserving chemical behavior of the parent compound, enabling accurate quantification and tracer studies in mass spectrometry and related assays.
  • Deuterium-labeled reference standard for LC-MS analyses.
  • Molecular weight 438.49 g/mol with defined chemical formula.
  • High purity suitable for quantitative analytical workflows.
  • Offered in small measured packages for easy sample preparation.
  • Datasheet and safety data sheet available for handling and characterization.