Learn More
Cayman Chemical OlaparIb-d4 500UG
SDP
Catalog No. 502893648
Encompass_Preferred
An internal standard for the quantification of olaparib by GC- or LC-MS
Catalog No. 50-289-3648 Supplier Cayman Chemical Supplier No. 9002700500 UG
An internal standard for the quantification of olaparib by GC- or LC-MS